Learn More
New England Biolabs, Inc. NdeI – 20000 units
SDP
Catalog No. 50811946
Encompass
A restriction endonuclease that recognizes the sequence CA^TA_TG.
Catalog No. 50-811-946 Supplier New England Biolabs, Inc. Supplier No. R0111L
A restriction endonuclease that recognizes the sequence CA^TA_TG.