Learn More
New England Biolabs, Inc. NdeI – 4000 units
SDP
Catalog No. 50811947
Encompass
A restriction endonuclease that recognizes the sequence CA^TA_TG.
Catalog No. 50-811-947 Supplier New England Biolabs, Inc. Supplier No. R0111S
A restriction endonuclease that recognizes the sequence CA^TA_TG.