Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ndelta-Allyloxycarbonyl-Nalpha-Fmoc-L-ornithine, 95%

Catalog No. AAH6333114
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6333114 25 g
AAH6333106 5 g
AAH6333103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6333114 Supplier Thermo Scientific Chemicals Supplier No. H6333114
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 147290-11-7 | C24H26N2O6 | 438.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147290-11-7
Molecular Formula C24H26N2O6
Molecular Weight (g/mol) 438.48
MDL Number MFCD00798637
InChI Key RXLIOYNXBHZZBI-NRFANRHFSA-N
Synonym fmoc-orn alloc-oh, fmoc-orn aloc-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy carbonyl amino pentanoic acid, n2-fmoc-n5-allyloxycarbonyl-l-ornithine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-prop-2-en-1-yloxy carbonyl amino pentanoic acid, pubchem6308, n, a-alloc-n, a-fmoc-l-ornithine, a-fmoc-n, a-alloc-l-ornithine, n-a-boc-n-d-allyloxycarbonyl-l-ornithine, n-alpha-9-fluorenylmethyloxycarbonyl-n-delta-allyloxycarbonyl-l-ornithine
PubChem CID 2756120
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-(prop-2-enoxycarbonylamino)pentanoic acid
SMILES C=CCOC(=O)NCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Quantity 25 g
Formula Weight 438.47
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Ndelta-Allyloxycarbonyl-Nalpha-Fmoc-L-ornithine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.