Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ndelta-Boc-Nalpha-Fmoc-D-ornithine, 95%

Catalog No. AAH6293003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6293003 1 g
AAH6293006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6293003 Supplier Thermo Scientific Chemicals Supplier No. H6293003
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 118476-89-4 | C25H30N2O6 | 454.523 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118476-89-4
Molecular Formula C25H30N2O6
Molecular Weight (g/mol) 454.523
MDL Number MFCD00077065
InChI Key JOOIZTMAHNLNHE-OAQYLSRUSA-N
Synonym fmoc-d-orn boc-oh, n-fmoc-n'-boc-d-ornithine, 2r-5-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, r-2-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxycarbonyl amino pentanoic acid, fmoc-n'-boc-d-ornithine, ambotzfaa1492, fmoc-nd-boc-d-ornithine, n-alpha-fmoc-nd-t-boc-d-ornithine, r-n5-tert.-butoxy-carbonyl-n2-9-fluorenylmethoxycarbonyl-ornithine, 2r-5-tert-butoxycarbonylamino-2-9h-fluoren-9-ylmethoxycarbonylamino pentanoic acid
PubChem CID 13996154
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC(C)(C)OC(=O)NCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Color Yellow
Quantity 1 g
Formula Weight 454.52
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Ndelta-Boc-Nalpha-Fmoc-D-ornithine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.