Learn More
Cayman Chemical NDemethylvncomycInhydroch 10mg
SDP
Catalog No. 502947828
Encompass_Preferred
A glycopeptide antibiotic; active against several strains of S. aureus and S. epidermidis in vitro and in vivo
Catalog No. 50-294-7828 Supplier Cayman Chemical Supplier No. 3411910 MG
A glycopeptide antibiotic; active against several strains of S. aureus and S. epidermidis in vitro and in vivo