Learn More
Cayman Chemical N-Demethylvancomycin (hydrochloride), Purity: ≥95%, CAS: 198774-23-1, 50 mg
SDP
Catalog No. 502949170
Encompass_Preferred
A glycopeptide antibiotic; active against several strains of S. aureus and S. epidermidis in vitro and in vivo
Catalog No. 50-294-9170 Supplier Cayman Chemical Supplier No. 3411950 MG
A glycopeptide antibiotic; active against several strains of S. aureus and S. epidermidis in vitro and in vivo