Learn More
Cayman Chemical NDesethylamIodaronhydroch 10mg
SDP
Catalog No. 502894346
Encompass_Preferred
the major CYP3A4 metabolite of amiodarone; used as an analytical reference standard for monitoring clinical efficacy of amiodarone in plasma samples
Catalog No. 50-289-4346 Supplier Cayman Chemical Supplier No. 900053710 MG
the major CYP3A4 metabolite of amiodarone; used as an analytical reference standard for monitoring clinical efficacy of amiodarone in plasma samples