Learn More
Cayman Chemical N-Desethylamiodarone (hydrochloride), Purity: ≥95%, CAS: 96027-74-6, 50 mg
SDP
Catalog No. 502894228
Encompass_Preferred
the major CYP3A4 metabolite of amiodarone; used as an analytical reference standard for monitoring clinical efficacy of amiodarone in plasma samples
Catalog No. 50-289-4228 Supplier Cayman Chemical Supplier No. 900053750 MG
the major CYP3A4 metabolite of amiodarone; used as an analytical reference standard for monitoring clinical efficacy of amiodarone in plasma samples