Learn More
Cayman Chemical N-desmethyl Sildenafil (citrate), Purity: ≥98%, CAS: 2319839-04-6, 5 mg
SDP
Catalog No. 502891425
Encompass_Preferred
A major metabolite of sildenafil; formed via oxidative metabolism of sildenafil by CYP3A4, CYP3A5, and CYP3A7
Catalog No. 50-289-1425 Supplier Cayman Chemical Supplier No. 100105875 MG
A major metabolite of sildenafil; formed via oxidative metabolism of sildenafil by CYP3A4, CYP3A5, and CYP3A7