Learn More
Cayman Chemical Ndesmethyl Sildenafil citrat
SDP
Supplier:  Cayman Chemical 100105871 MG
Encompass_Preferred
A major metabolite of sildenafil; formed via oxidative metabolism of sildenafil by CYP3A4, CYP3A5, and CYP3A7
Catalog No. 50-288-7126