Learn More
Med Vet International BD 18G x 1.5 in. Blunt Fill Needles, 100/box
Catalog No. 503045029
Encompass_Preferred
This BD 305180 Needle is a 18G x 1.5" Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is a blunt filled needle. It is designed for vial access. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.
Catalog No. 50-304-5029 Supplier Med Vet International Supplier No. 305180
This BD 305180 Needle is a 18G x 1.5" Regular Wall needle. Regular wall needles are more rigid than thin wall needles, providing extra support and less flex due to added annula thickness. This is a blunt filled needle. It is designed for vial access. NY, CT, IL, NJ, NH, DE and MN restrict the sale of syringes and needles to licensed professionals and entities. By proceeding to purchase through our website, you acknowledge that you are over 18 years of age.