Learn More
SAI Infusion Technologies 20 Gauge, 0.5" Blunt Needles - Non-sterile, 100/pk
SDP
Catalog No. NC0183331
Encompass
20G0.5" Non-Sterile Blunt Needles with plastic hub and stainless steel blunt cannula. These needles are non-sterile, in a bulk case of 100. ETO Exposed sterile blunt needles are available separately. Product is for laboratory animal research only.
Catalog No. NC0183331 Supplier SAI Infusion Technologies Supplier No. B2050 100 BULK
20G0.5" Non-Sterile Blunt Needles with plastic hub and stainless steel blunt cannula. These needles are non-sterile, in a bulk case of 100. ETO Exposed sterile blunt needles are available separately. Product is for laboratory animal research only.