Learn More
Med Vet International BD 305761 - Eclipse Safety Needle 21g X 1", 100/Box
Catalog No. 503044911
Encompass_Preferred
The BD Eclipse needle is a pivoting-shield needle for efficient safety during activation. The needle is bevel oriented to the safety cover to facilitate low-angle injections.Features and benefitsThe BD Eclipse needle provides streamlined and effective injection safety.Activation that requires virtually no change in clinical technique for easy integration.Designed to specifically reduce the risk of accidental needlestick injuries.Allows for shielding of the needle tip immediately after injection, with a single finger stroke.Audible click confirms activation to support your safety.
Catalog No. 50-304-4911 Supplier Med Vet International Supplier No. 305764
The BD Eclipse needle is a pivoting-shield needle for efficient safety during activation. The needle is bevel oriented to the safety cover to facilitate low-angle injections.Features and benefitsThe BD Eclipse needle provides streamlined and effective injection safety.Activation that requires virtually no change in clinical technique for easy integration.Designed to specifically reduce the risk of accidental needlestick injuries.Allows for shielding of the needle tip immediately after injection, with a single finger stroke.Audible click confirms activation to support your safety.