Learn More
Med Vet International BD Vacutainer Eclipse™ Blood Collection Needle 21 Gauge 1-1/4 Inch Needle Length Safety Needle Without Tubing Sterile, 368607, 480/Case
Catalog No. 503044867
Encompass_Preferred
Single handed activation Integrated safety shield that fully covers needle after use Unique double locking system Audible click indicator
Catalog No. 50-304-4867 Supplier Med Vet International Supplier No. 373902CS
Single handed activation Integrated safety shield that fully covers needle after use Unique double locking system Audible click indicator