Learn More
Med Vet International BD Vacutainer Eclipse™ Blood Collection Needle 22 Gauge 1-1/4 Inch Needle Length Safety Needle Without Tubing Sterile, 368608, 480/Case
Catalog No. 503043118
Encompass_Preferred
Single handed activation Integrated safety shield that fully covers needle after use Unique double locking system Audible click indicator
Catalog No. 50-304-3118 Supplier Med Vet International Supplier No. 373903CS
Single handed activation Integrated safety shield that fully covers needle after use Unique double locking system Audible click indicator