Learn More
Gerstel Gerstel Replacement Needle, Wide Bore, Gastight Performance, 2 per Pack
SDP
Catalog No. NC0987517
Encompass
The GERSTEL Replacement Needle is designed for 1.0 and 2.5 mL TriStar Headspace Syringes, offering a wide bore for enhanced analysis. Crafted from rugged, inert materials, it minimizes carry-over and resists various solvents, ensuring long-lasting, gas-tight performance crucial in chromatographic applications. With two needles per pack, it provides an efficient solution for demanding laboratory needs.
Catalog No. NC0987517 Supplier Gerstel Supplier No. 02200180300
Add to Cart
Add to Cart
The GERSTEL Replacement Needle is designed for 1.0 and 2.5 mL TriStar Headspace Syringes, offering a wide bore for enhanced analysis. Crafted from rugged, inert materials, it minimizes carry-over and resists various solvents, ensuring long-lasting, gas-tight performance crucial in chromatographic applications. With two needles per pack, it provides an efficient solution for demanding laboratory needs.