Learn More
Med Vet International Olsen Hegar Needle Holder, 6.5", Economy, Satin, Each
Supplier:  Med Vet International 111235S
Encompass_Preferred
Olsen-Hegar Needle Holders are a ratcheted, finger ring instrument that also has built in scissors. It is used with intermediate weight sutures, commonly in plastic surgery procedures. This product is straight with delicate, cross-serrated tips.
Catalog No. 50-222-3965