Learn More
Med Vet International AHS Needle, 22 X 3/4", Hypodermic, 1000/BX, AHS2219RN
Catalog No. 503045512
Encompass_Preferred
This AHS AHS2219RN Needle is a 22 X 3/4" Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 1000/BX
Catalog No. 50-304-5512 Supplier Med Vet International Supplier No. AHS2219RNX10
This AHS AHS2219RN Needle is a 22 X 3/4" Thin Wall needle. Thin wall needles are thinner than regular wall needles. The outside diameter is the same size. However, the inside diameter is bigger than the regular wall's inside diameter. Thin wall needles provide more comfort with an increased flow rate. This hypodermic needle is a hollow needle used for withdrawing fluid like blood samples, or injecting medications. This quantity is 1000/BX