Learn More
Trajan Scientific NDL N10-VA8000H-II 0.20
SDP
Catalog No. 502530263
Encompass_Preferred
Trajan Varian/Bruker Replacement Needle, Diameter Outer: 0.5 mm, Needle Gauge: 25 G, Length Needle: 53 mm, Needle Point Style: S-hole, For Use With: For autosampler syringes, 8035, 8100 and 8200, Replacement Needle Varian/Bruker
Catalog No. 50-253-0263 Supplier Trajan Scientific Supplier No. 037780
Trajan Varian/Bruker Replacement Needle, Diameter Outer: 0.5 mm, Needle Gauge: 25 G, Length Needle: 53 mm, Needle Point Style: S-hole, For Use With: For autosampler syringes, 8035, 8100 and 8200, Replacement Needle Varian/Bruker