Learn More
Trajan Scientific NDL NM5/10-LC 5PK
SDP
Catalog No. 502530288
Encompass_Preferred
Trajan Replacement Needle for 5 and 10mL Syringes, Diameter Inner: 0.014 in., 0.37 mm, Needle Gauge: 22 G, Length Needle: 2 in., 51 mm, Needle Point Style: Removable Rheodyne, Diameter Outer: 0.028 in., 0.63 mm, For Use With: 5 and 10mL syringe, Stainless Steel
Catalog No. 50-253-0288 Supplier Trajan Scientific Supplier No. 0315233
Trajan Replacement Needle for 5 and 10mL Syringes, Diameter Inner: 0.014 in., 0.37 mm, Needle Gauge: 22 G, Length Needle: 2 in., 51 mm, Needle Point Style: Removable Rheodyne, Diameter Outer: 0.028 in., 0.63 mm, For Use With: 5 and 10mL syringe, Stainless Steel