Learn More
Trajan Scientific NDL NP1B-7/BV
SDP
Catalog No. 502530268
Encompass_Preferred
SGE Replacement Needles for 0.5 - 1uL Syringes, For Use With: For 8mm O.D. barrel 1uL microvolume syringes, Needle Gauge: 23 G, Length Needle: 2.7 in., 70 mm, Needle Point Style: Bevel, Diameter Inner: 0.006 in., 0.15 mm, Diameter Outer: 0.02 in., 0.5 mm, Stainless Steel
Catalog No. 50-253-0268 Supplier Trajan Scientific Supplier No. 034060
SGE Replacement Needles for 0.5 - 1uL Syringes, For Use With: For 8mm O.D. barrel 1uL microvolume syringes, Needle Gauge: 23 G, Length Needle: 2.7 in., 70 mm, Needle Point Style: Bevel, Diameter Inner: 0.006 in., 0.15 mm, Diameter Outer: 0.02 in., 0.5 mm, Stainless Steel