Learn More
Gerstel NDL NRW BR .15MM X 57MM LG 2PK
SDP
Catalog No. NC0968534
Encompass
GERSTEL TriStar HS syringes for Headspace injection up to 100�C. Replacement needle. Volume 1-2.5mL. OD/ID/L/Tip [mm] 0.63/0.15/57.0/HS. 2 each per pack.
Catalog No. NC0968534 Supplier Gerstel Supplier No. 02200180200
GERSTEL TriStar HS syringes for Headspace injection up to 100�C. Replacement needle. Volume 1-2.5mL. OD/ID/L/Tip [mm] 0.63/0.15/57.0/HS. 2 each per pack.