Learn More
Trajan Scientific NDL P50-GT
SDP
Catalog No. 502530279
Encompass_Preferred
Trajan Replacement Plungers, For Use With: For Rheodyne and Valco valves, 50 mL Replacement Plunger only, Length Needle: 50.8 mm, 2 in., Needle Gauge: 22 G, Needle Point Style: Removable LC, Volume: 50 uL, Diameter Outer Needle: 0.028 in., 0.71 mm, Length: 2 in., 51 mm, Model: 50R-GT-LC
Catalog No. 50-253-0279 Supplier Trajan Scientific Supplier No. 031820
Trajan Replacement Plungers, For Use With: For Rheodyne and Valco valves, 50 mL Replacement Plunger only, Length Needle: 50.8 mm, 2 in., Needle Gauge: 22 G, Needle Point Style: Removable LC, Volume: 50 uL, Diameter Outer Needle: 0.028 in., 0.71 mm, Length: 2 in., 51 mm, Model: 50R-GT-LC