Learn More
Med Vet International McKesson Hypodermic Needle, 31 Gauge, 1/4 Inch Length, 16-N6MMPA, 2000/Case
Catalog No. 503043738
Encompass_Preferred
Insulin Pen Needle McKesson Prevent 31 Gauge 1/4 Inch Length Safety Shield Automatic needle lock-out; appearance of red stripes confirms that the needle has been used and is ready to be disposed of safely Reduces user discomfort due to thinner needle (31G) Viewing window allows easy visibility to needle tip during priming to prevent unnecessary drug waste Wing-shaped design for comfortable and secure grip Regular bevel Stainless steel needle is a 1/4 inch in length
Catalog No. 50-304-3738 Supplier Med Vet International Supplier No. 1118151CS
Insulin Pen Needle McKesson Prevent 31 Gauge 1/4 Inch Length Safety Shield Automatic needle lock-out; appearance of red stripes confirms that the needle has been used and is ready to be disposed of safely Reduces user discomfort due to thinner needle (31G) Viewing window allows easy visibility to needle tip during priming to prevent unnecessary drug waste Wing-shaped design for comfortable and secure grip Regular bevel Stainless steel needle is a 1/4 inch in length