Learn More
Trajan Scientific NDL SER II NLL-11.5/14 5/PK
SDP
Catalog No. 502530250
Encompass_Preferred
Trajan Replaceable Luer Lock Needles: Bevel, Length Needle: 115 mm, 4.52 in., Needle Gauge: 14 G, Needle Point Style: Bevel, Diameter Inner: 0.062 in., 1.6 mm, Diameter Outer: 0.062 in., 2.11 mm, For Use With: Gas chromatography, Stainless Steel, Termination: Luer-lock, Replacement Needle
Catalog No. 50-253-0250 Supplier Trajan Scientific Supplier No. 039891
Trajan Replaceable Luer Lock Needles: Bevel, Length Needle: 115 mm, 4.52 in., Needle Gauge: 14 G, Needle Point Style: Bevel, Diameter Inner: 0.062 in., 1.6 mm, Diameter Outer: 0.062 in., 2.11 mm, For Use With: Gas chromatography, Stainless Steel, Termination: Luer-lock, Replacement Needle