Learn More
Med Vet International Quincke Spinal Needle, 20ga x 2-1/2in., 1/Pkg
Catalog No. 503044830
Encompass_Preferred
Sterile, single use. Key/slot arrangement of stylet and cannula hubs facilitates proper needle bevel orientation. Translucent window hub features contact clarity that helps allow visualization of CSF. Specialized fitted stylet reduces tissue coring
Catalog No. 50-304-4830 Supplier Med Vet International Supplier No. 405071
Sterile, single use. Key/slot arrangement of stylet and cannula hubs facilitates proper needle bevel orientation. Translucent window hub features contact clarity that helps allow visualization of CSF. Specialized fitted stylet reduces tissue coring