Learn More
Med Vet International BD Spinal Needle Yale, 22G x 1-1/2", 25/Box
Catalog No. 503043122
Encompass_Preferred
Polypropylene Hub Key/Slot arrangement of stylet and cannula hubs facilitates proper needle bevel orientation Unique translucent window hub allows for easy visualization of CSF Specialized fitted stylet reduces tissue coring
Catalog No. 50-304-3122 Supplier Med Vet International Supplier No. 405161
Polypropylene Hub Key/Slot arrangement of stylet and cannula hubs facilitates proper needle bevel orientation Unique translucent window hub allows for easy visualization of CSF Specialized fitted stylet reduces tissue coring