Learn More
Trajan Scientific NDL SW10 5PK
SDP
Catalog No. 502530287
Encompass_Preferred
Trajan Stylet Wires for Syringe Cleaning, For Use With: 5 to 10uL syringe needles, Replacement Stylet Wire, Length: 5.9 in., 15 cm
Catalog No. 50-253-0287 Supplier Trajan Scientific Supplier No. 031745
Trajan Stylet Wires for Syringe Cleaning, For Use With: 5 to 10uL syringe needles, Replacement Stylet Wire, Length: 5.9 in., 15 cm