Learn More
Trajan Scientific NDL SW15 5PK
SDP
Catalog No. 502530286
Encompass_Preferred
Trajan Stylet Wires for Syringe Cleaning, For Use With: 15 to 500uL syringe needles, Replacement Stylet Wire, Length: 5.9 in., 15 cm
Catalog No. 50-253-0286 Supplier Trajan Scientific Supplier No. 031746
Trajan Stylet Wires for Syringe Cleaning, For Use With: 15 to 500uL syringe needles, Replacement Stylet Wire, Length: 5.9 in., 15 cm