Learn More
Med Vet International Exel 17g x 1.5in. Blunt Aluminum Hub Needle, 25/box
Supplier:  Med Vet International 26472
Encompass_Preferred
The Exel International 26472 aluminum hub blunt needles have a luer lock design and are sterile, single use, and individually packaged. They have a stainless-steel laser welded non lubricated cannula and have a 2 piece polypropyleneneedle cap and sheath.
Catalog No. 50-255-9742