Learn More
Med Vet International Exel 18g x 1in. Blunt Aluminum Hub Needle, 25/box
Catalog No. 502559741
Encompass_Preferred
The Exel International 26473 aluminum hub blunt needles have a luer lock design and are sterile, single use, and individually packaged. They have a stainless-steel laser welded non lubricated cannula and have a 2 piece polypropyleneneedle cap and sheath.
Catalog No. 50-255-9741 Supplier Med Vet International Supplier No. 26473
The Exel International 26473 aluminum hub blunt needles have a luer lock design and are sterile, single use, and individually packaged. They have a stainless-steel laser welded non lubricated cannula and have a 2 piece polypropyleneneedle cap and sheath.