Learn More
Med Vet International EasyTouch Pen Needles, 30G x 5/16in, 100/Box
Catalog No. 502559637
Encompass_Preferred
Each EasyTouch Pen Needle cap is color-coded by gauge for easy and accurate identification. Confusion is reduced significantly by allowing customers to quickly identify their correct gauge and needle length. Color-Coded to GaugeEasy-to-read information on each side of the box. Needles are made with Surgical Steel. Triple-Bevel Cut for a Sharper Point. Thin Wall Cannulas for an Easier Flow. Electro-polished to Remove Burrs for a Smoother InjectionFilm Coated for Maximum Comfort
Catalog No. 50-255-9637 Supplier Med Vet International Supplier No. 830561
Each EasyTouch Pen Needle cap is color-coded by gauge for easy and accurate identification. Confusion is reduced significantly by allowing customers to quickly identify their correct gauge and needle length. Color-Coded to GaugeEasy-to-read information on each side of the box. Needles are made with Surgical Steel. Triple-Bevel Cut for a Sharper Point. Thin Wall Cannulas for an Easier Flow. Electro-polished to Remove Burrs for a Smoother InjectionFilm Coated for Maximum Comfort