Learn More
Med Vet International EasyTouch Pen Needles, 31G x 5/16in, 100/Box
Supplier:  Med Vet International 831061
Encompass_Preferred
Each EasyTouch Pen Needle cap is color-coded by gauge for easy and accurate identification. Confusion is reduced significantly by allowing customers to quickly identify their correct gauge and needle length. Color-Coded to Gauge Easy-to-read information on each side of the box Needles are made with Surgical Steel Triple-Bevel Cut for a Sharper Point Thin Wall Cannulas for an Easier Flow Electro-polished to Remove Burrs for a Smoother Injection Film Coated for Maximum Comfort
Catalog No. 50-255-9634