Learn More
Med Vet International EasyTouch Safety Pen Needles, 30G x 3/16in, 100/Box
Catalog No. 502559639
Encompass_Preferred
Easy Touch Safety Pen Needles feature a protective shield that automatically locks into place after injection to help reduce the occurrence of neddlestick injuries. Designed and manufactured to the highest quality levels possible the Easy Touch safety pen needles are film coated for maximum comfort. You will quickly recognize this level of quality as you experience a more comfortable and safer injection.
Catalog No. 50-255-9639 Supplier Med Vet International Supplier No. 830536
Easy Touch Safety Pen Needles feature a protective shield that automatically locks into place after injection to help reduce the occurrence of neddlestick injuries. Designed and manufactured to the highest quality levels possible the Easy Touch safety pen needles are film coated for maximum comfort. You will quickly recognize this level of quality as you experience a more comfortable and safer injection.