Learn More
Aviva Systems Biology Corp NDN Antibody - middle region (ARP56358_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56358_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NDN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NDN. Alternate names: HsT16328, PWCRApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-61828