Learn More
Aviva Systems Biology Corp NSMCE3 Antibody - middle region (ARP57727_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57727_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NDNL2. It was validated on Western Blot. NDNL2,necdin-like 2,HCA4; NSE3; MAGEG1; MAGEL3; NSMCE3; NDNL2,melanoma-associated antigen G1,melanoma-associated antigen G1,MAGE-G1 antigen,necdin-like gene 2,necdin-like protein 2,melanoma antigen, family G, 1,hepatocellular carcinoma-associated protein 4,hepatocellular carcinoma-associated protein HCA4The immunogen is a synthetic peptide directed towards the middle region of human NDNL2. Alternate names: HCA4, MAGEG1, MAGEL3, NSE3, NSMCE3ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-365