Learn More
Cayman Chemical ndocromIlsodIum salt 50mg
SDP
Catalog No. 502933091
Encompass_Preferred
A mast cell stabilizer; inhibits IgE-induced histamine release from MC/3T3 cells preincubated with IL-2 at 10 µM; prevents EArly- and late-phase bronchoconstriction in a guinea pig model of asthma induced by ovalbumin; prevents conjunctival edema and erythema and reduces mast cell degranulation in a rat model of conjunctivitis at 2 mg/kg
Catalog No. 50-293-3091 Supplier Cayman Chemical Supplier No. 2400550 MG
A mast cell stabilizer; inhibits IgE-induced histamine release from MC/3T3 cells preincubated with IL-2 at 10 µM; prevents EArly- and late-phase bronchoconstriction in a guinea pig model of asthma induced by ovalbumin; prevents conjunctival edema and erythema and reduces mast cell degranulation in a rat model of conjunctivitis at 2 mg/kg