Learn More
Cayman Chemical NdodecnoylLHomoserIn lac 10mg
SDP
Catalog No. 502893551
Encompass_Preferred
A fluorescently tagged probe which allows the HSL molecule to be detected using fluorescence-based techniques
Catalog No. 50-289-3551 Supplier Cayman Chemical Supplier No. 900071110 MG
A fluorescently tagged probe which allows the HSL molecule to be detected using fluorescence-based techniques