Learn More
Aviva Systems Biology Corp NDRG1 Antibody - C-terminal region (ARP47974_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47974_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NDRG1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NDRG1. Alternate names: CAP43, CMT4D, DRG1, GC4, HMSNL, NDR1, NMSL, PROXY1, RIT42, RTP, TARG1, TDD5ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59691