Learn More
Aviva Systems Biology Corp NDRG2 Antibody - C-terminal region (ARP47976_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47976_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NDRG2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NDRG2. Alternate names: DKFZp781G1938, FLJ25522, KIAA1248, SYLDApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59693