Learn More
Aviva Systems Biology Corp Ndrg2 Antibody - middle region (ARP47977_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47977_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ndrg2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AI182517, AU040374, Ndr2, SYLDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-336