Learn More
Aviva Systems Biology Corp NDRG4 Antibody - N-terminal region (ARP64736_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64736_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NDRG4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human NDRG4. Alternate names: DKFZp686I1615, FLJ30586, FLJ42011, KIAA1180, MGC19632, SMAP-8ApplicationWB. Species reactivity: Human, Pig, Rat
Catalog No. 50-288-350