Learn More
Cayman Chemical Androgen Receptor Bioassay, 96 Well
SDP
Catalog No. 502942889
Encompass_Preferred
In addition to AR Reporter Cells, this kit includes two optimized media for use during cellculture and in diluting the user's test samples, Luciferase Detection Reagent, and a cell culture-ready assay plate.
Catalog No. 50-294-2889 Supplier Cayman Chemical Supplier No. 4103696 WELL
In addition to AR Reporter Cells, this kit includes two optimized media for use during cellculture and in diluting the user's test samples, Luciferase Detection Reagent, and a cell culture-ready assay plate.