Learn More
Cayman Chemical Andrographolide, Purity: ≥98%, CAS: 5508-58-7, 100 mg
SDP
Catalog No. 502952383
Encompass_Preferred
A diterpenoid compound that inhibits NF-κB binding to DNA promoters and induces expression of the CYP1A enzymes to produce immunosuppressant, antithrombotic, anti-inflammatory, antineoplastic, antiviral, antibacterial, antidiabetic, antioxidative stress, antipyretic, antiedematogenic, and antinociceptive activities
Catalog No. 50-295-2383 Supplier Cayman Chemical Supplier No. 11679100 MG
A diterpenoid compound that inhibits NF-κB binding to DNA promoters and induces expression of the CYP1A enzymes to produce immunosuppressant, antithrombotic, anti-inflammatory, antineoplastic, antiviral, antibacterial, antidiabetic, antioxidative stress, antipyretic, antiedematogenic, and antinociceptive activities