Learn More
Cayman Chemical ndrost-3 5-dIen-7 17-dIon 10mg
SDP
Catalog No. 502964357
Encompass_Preferred
An analytical reference standard categorized as an androgenic anabolic steroid; a metabolite of 7-keto-dehydroepiandrosterone; intended for research and forensic applications
Catalog No. 50-296-4357 Supplier Cayman Chemical Supplier No. 2308610 MG
An analytical reference standard categorized as an androgenic anabolic steroid; a metabolite of 7-keto-dehydroepiandrosterone; intended for research and forensic applications