Learn More
Cayman Chemical Androsterone, Purity: ≥98%, CAS: 53-41-8, 100 mg
SDP
Catalog No. 502971793
Encompass_Preferred
A steroid hormone produced in the body from 5α-reduced metabolites of dehydroepiandrosterone that possesses 7-fold weaker androgen potency compared to that of testosterone; excreted in urine and used as the international reference standard for androgenic activity
Catalog No. 50-297-1793 Supplier Cayman Chemical Supplier No. 15872100 MG
A steroid hormone produced in the body from 5α-reduced metabolites of dehydroepiandrosterone that possesses 7-fold weaker androgen potency compared to that of testosterone; excreted in urine and used as the international reference standard for androgenic activity