Learn More
Hampton Research Non-detergent sulfobetaine. Crystallization grade NDSB-195 5gr
SDP
Catalog No. 502711
Encompass
Non-detergent sulfobetaine. Crystallization grade NDSB-195 for formulating screens or for optimization. Please note items from this supplier are non-returnable.
Catalog No. 50-271-1 Supplier Hampton Research Supplier No. HR2703
Non-detergent sulfobetaine. Crystallization grade NDSB-195 for formulating screens or for optimization. Please note items from this supplier are non-returnable.