Learn More
Gold Biotechnology Inc NDSB-201 (Non-detergent sulfobetaine) 100 g
SDP
Catalog No. NC1517081
Encompass
NDSB-201 is a protein specific additive that can be used to help solubilize and stabilize proteins at non-denaturing concentrations while preventing protein aggregation.
Catalog No. NC1517081 Supplier Gold Biotechnology Inc Supplier No. N201100
NDSB-201 is a protein specific additive that can be used to help solubilize and stabilize proteins at non-denaturing concentrations while preventing protein aggregation.