Learn More
Hampton Research Non-detergent sulfobetaine. Crystallization grade NDSB-221 5gr
SDP
Catalog No. 502713
Encompass
Non-detergent sulfobetaine. Crystallization grade NDSB-221 for formulating screens or for optimization. Please note items from this supplier are non-returnable.
Catalog No. 50-271-3 Supplier Hampton Research Supplier No. HR2791
Non-detergent sulfobetaine. Crystallization grade NDSB-221 for formulating screens or for optimization. Please note items from this supplier are non-returnable.