Learn More
Cayman Chemical N-desethyl Brinzolamide (oxalate), Purity: ≥95%, CAS: 2775292-24-3, 10 mg
SDP
Catalog No. 502950137
Encompass_Preferred
An active metabolite of brinzolamide; inhibits CAII and CAIV activities (IC50s = 1.28 and 128 nM, respectively)
Catalog No. 50-295-0137 Supplier Cayman Chemical Supplier No. 3427510 MG
An active metabolite of brinzolamide; inhibits CAII and CAIV activities (IC50s = 1.28 and 128 nM, respectively)